siRNA Details
| Gene Symbol: Thap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008340.1 | NP_001008341.1 | 1 | 277 | ||
| GenBank mRNA | BC086347.1 | - | 1 | 277 | 7302510 | |
| JF423321.1 | - | 1 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thap1| Gene Symbol: Thap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008340.1 | NP_001008341.1 | 1 | 277 | ||
| GenBank mRNA | BC086347.1 | - | 1 | 277 | 7302510 | |
| JF423321.1 | - | 1 | 277 | |||