siRNA Details
Gene Symbol: RGD1309594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008351.1 | NP_001008352.1 | 2, 3 | 143 | ||
GenBank mRNA | BC085944.1 | - | 2, 3 | 143 | 7127440 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1309594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008351.1 | NP_001008352.1 | 2, 3 | 143 | ||
GenBank mRNA | BC085944.1 | - | 2, 3 | 143 | 7127440 |