siRNA Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008352.1 | NP_001008353.1 | 1, 2 | 148 | ||
| GenBank mRNA | BC086349.1 | - | 1, 2 | 148 | 7302798 | |
| FQ220630.1 | - | 1, 2 | 198 | |||
| FQ233923.1 | - | 1, 2 | 278 | |||
| FQ234368.1 | - | 1, 2 | 278 | |||

Pmvk
Genomic Map