siRNA Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008352.1 | NP_001008353.1 | 5 | 840 | ||
| GenBank mRNA | BC086349.1 | - | 5 | 840 | 7302798 | |
| FQ226470.1 | - | 5 | 970 | |||
| FQ233923.1 | - | 5 | 970 | |||
| FQ234368.1 | - | 5 | 970 | |||

Pmvk
Genomic Map