siRNA Details
| Gene Symbol: Mkks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008353.2 | NP_001008354.1 | 3 | 1537 | ||
| XM_008762254.1 | XP_008760476.1 | 3 | 1497 | |||
| GenBank mRNA | BC083863.1 | - | 3 | 1515 | 7127425 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkks| Gene Symbol: Mkks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008353.2 | NP_001008354.1 | 3 | 1537 | ||
| XM_008762254.1 | XP_008760476.1 | 3 | 1497 | |||
| GenBank mRNA | BC083863.1 | - | 3 | 1515 | 7127425 | |