siRNA Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008355.1 | NP_001008356.1 | 8 | 974 | ||
| GenBank mRNA | BC085846.1 | - | 8 | 974 | 7189020 | |
| FQ220446.1 | - | 8 | 859 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc3| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008355.1 | NP_001008356.1 | 8 | 974 | ||
| GenBank mRNA | BC085846.1 | - | 8 | 974 | 7189020 | |
| FQ220446.1 | - | 8 | 859 | |||