siRNA Details
| Gene Symbol: Hcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008357.1 | NP_001008358.1 | 7 | 1058 | ||
| GenBank mRNA | BC085951.1 | - | 7 | 1058 | 7132601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcfc2| Gene Symbol: Hcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008357.1 | NP_001008358.1 | 7 | 1058 | ||
| GenBank mRNA | BC085951.1 | - | 7 | 1058 | 7132601 | |