siRNA Details
| Gene Symbol: Cdip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008360.1 | NP_001008361.1 | 7 | 1747 | ||
| XM_006245811.2 | XP_006245873.1 | 6 | 1758 | |||
| XM_006245812.2 | XP_006245874.1 | 8 | 1945 | |||
| XM_006245813.2 | XP_006245875.1 | 7 | 2268 | |||
| XM_006245814.2 | XP_006245876.1 | 7 | 1871 | |||
| XM_006245815.2 | XP_006245877.1 | 6 | 2194 | |||
| XM_006245816.2 | XP_006245878.1 | 6 | 1761 | |||
| XM_008767493.1 | XP_008765715.1 | 6 | 3172 | |||
| XM_008767494.1 | XP_008765716.1 | 6 | 3423 | |||
| GenBank mRNA | BC085860.1 | - | 7 | 1747 | 7190340 | |

Cdip1
Genomic Map