siRNA Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008363.1 | NP_001008364.1 | 7 | 1120 | ||
| GenBank mRNA | BC085928.1 | - | 7 | 1120 | 7110398 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfand2a| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008363.1 | NP_001008364.1 | 7 | 1120 | ||
| GenBank mRNA | BC085928.1 | - | 7 | 1120 | 7110398 | |