siRNA Details
| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008364.1 | NP_001008365.1 | 7 | 1457 | ||
| XM_008772152.1 | XP_008770374.1 | 8 | 1504 | |||
| GenBank mRNA | BC085883.1 | - | 7 | 1457 | 7192093 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx24| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008364.1 | NP_001008365.1 | 7 | 1457 | ||
| XM_008772152.1 | XP_008770374.1 | 8 | 1504 | |||
| GenBank mRNA | BC085883.1 | - | 7 | 1457 | 7192093 | |