siRNA Details
| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008365.1 | NP_001008366.1 | 5 | 433 | ||
| XM_006255145.2 | XP_006255207.1 | 5 | 525 | |||
| GenBank mRNA | BC083873.1 | - | 5 | 433 | 7128979 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tom1| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008365.1 | NP_001008366.1 | 5 | 433 | ||
| XM_006255145.2 | XP_006255207.1 | 5 | 525 | |||
| GenBank mRNA | BC083873.1 | - | 5 | 433 | 7128979 | |