siRNA Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008366.1 | NP_001008367.1 | 6 | 517 | ||
| XM_006255686.2 | XP_006255748.1 | 6 | 631 | |||
| XM_006255687.2 | XP_006255749.1 | 6 | 549 | |||
| XM_006255689.2 | XP_006255751.1 | 7 | 853 | |||
| XM_006255690.2 | XP_006255752.1 | 7 | 639 | |||
| XM_006255691.2 | XP_006255753.1 | 5 | 573 | |||
| XM_006255692.2 | XP_006255754.1 | 5 | 453 | |||
| GenBank mRNA | BC085839.1 | - | 6 | 517 | 7135799 | |

Cenpn
Genomic Map