siRNA Details
| Gene Symbol: CROCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014675.4 | NP_055490.4 | 27 | 4136 | ||
| XM_006711058.3 | XP_006711121.1 | 26 | 3927 | |||
| XM_011542468.1 | XP_011540770.1 | 27 | 4113 | |||
| XM_011542470.2 | XP_011540772.1 | 27 | 4113 | |||
| XM_017002913.1 | XP_016858402.1 | 27 | 4113 | |||
| XM_017002914.1 | XP_016858403.1 | 27 | 4113 | |||
| XM_017002915.1 | XP_016858404.1 | 24 | 3621 | |||
| GenBank mRNA | AB007914.2 | - | 25 | 3775 | ||
| DQ139275.1 | - | 14 | 3493 | |||

CROCC
Genomic Map