siRNA Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008372.1 | NP_001008373.1 | 4 | 656 | ||
| XM_006231776.2 | XP_006231838.1 | 4 | 646 | |||
| XM_006231778.2 | XP_006231840.1 | 4 | 695 | |||
| XM_008760639.1 | XP_008758861.1 | 5 | 759 | |||
| GenBank mRNA | BC085771.1 | - | 4 | 656 | 7191068 | |
| FQ211725.1 | - | 4 | 687 | |||

Papd4
Genomic Map