siRNA Details
| Gene Symbol: Ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008381.1 | NP_001008382.1 | 3, 4 | 215 | ||
| XM_008767275.1 | XP_008765497.1 | 3, 4 | 295 | |||
| GenBank mRNA | BC086355.1 | - | 3, 4 | 215 | 7303709 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2f| Gene Symbol: Ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008381.1 | NP_001008382.1 | 3, 4 | 215 | ||
| XM_008767275.1 | XP_008765497.1 | 3, 4 | 295 | |||
| GenBank mRNA | BC086355.1 | - | 3, 4 | 215 | 7303709 | |