siRNA Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008383.1 | NP_001008384.1 | 10 | 1586 | ||
| XM_006255517.2 | XP_006255579.1 | 11 | 1607 | |||
| XM_008772521.1 | XP_008770743.1 | 11 | 1804 | |||
| GenBank mRNA | BC085826.1 | - | 10 | 1586 | 7131855 | |

Dpep3
Genomic Map