siRNA Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008386.1 | NP_001008387.1 | 2 | 468 | ||
| XM_008764603.1 | XP_008762825.1 | 2 | 1819 | |||
| GenBank mRNA | AY769991.1 | - | 1 | 153 | ||
| AY769992.1 | - | 2 | 468 | |||

Ispd
Genomic Map