siRNA Details
| Gene Symbol: Gimap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008398.2 | NP_001008399.1 | 2 | 601 | ||
| GenBank mRNA | AJ633684.1 | - | 2 | 762 | ||
| BC103635.1 | - | 2 | 601 | 7100941 | ||
| DQ125337.1 | - | 2 | 600 | |||
| DQ125338.1 | - | 2 | 600 | |||

Gimap9
Genomic Map