siRNA Details
| Gene Symbol: C14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008404.2 | NP_001008404.1 | 3, 4 | 577 | ||
| NM_001286399.1 | NP_001273328.1 | 3, 4 | 577 | |||
| NM_001286400.1 | NP_001273329.1 | 3, 4 | 742 | |||
| XM_005267638.3 | XP_005267695.1 | 3, 4 | 690 | |||
| XM_011536764.2 | XP_011535066.1 | 3, 4 | 668 | |||
| GenBank mRNA | AK095913.1 | - | 3, 4 | 542 | ||
| AK297818.1 | - | 3, 4 | 497 | |||
| AK309489.1 | - | 3, 4 | 416 | |||
| BC041103.1 | - | 3, 4 | 480 | 5193008 | ||
| BX161457.1 | - | 3, 4 | 742 | |||

C14orf180
Genomic Map