siRNA Details
| Gene Symbol: KIAA0355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014686.3 | NP_055501.2 | 2 | 1184 | ||
| XM_005259444.1 | XP_005259501.1 | 2 | 1184 | |||
| XM_017027525.1 | XP_016883014.1 | 2 | 1184 | |||
| GenBank mRNA | AB002353.1 | - | 2 | 1163 | ||
| BC104761.1 | - | 1 | 345 | 8143764 | ||
| BC104763.1 | - | 1 | 345 | 8143766 | ||

KIAA0355
Genomic Map