siRNA Details
| Gene Symbol: ISX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303508.1 | NP_001290437.1 | 5 | 1387 | ||
| GenBank mRNA | AB219123.1 | - | 4 | 794 | ||
| AK025181.1 | - | 4 | 1676 | |||
| BC112212.1 | - | 4 | 785 | 8327680 | ||
| BC113633.1 | - | 4 | 773 | 8322685 | ||
| CR749372.1 | - | 5 | 1388 | |||

ISX
Genomic Map