siRNA Details
| Gene Symbol: Tpst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008508.2 | NP_001008508.1 | 3 | 1041 | ||
| XM_006249526.2 | XP_006249588.1 | 4 | 1121 | |||
| XM_008769319.1 | XP_008767541.1 | 4 | 1055 | |||
| GenBank mRNA | BC086518.1 | - | 3 | 1046 | 7302513 | |

Tpst2
Genomic Map