siRNA Details
| Gene Symbol: GPRIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014696.3 | NP_055511.2 | 3 | 1713 | ||
| XM_005277666.3 | XP_005277723.1 | 4 | 1989 | |||
| XM_011540388.2 | XP_011538690.1 | 6 | 2349 | |||
| XM_011540395.2 | XP_011538697.1 | 4 | 1957 | |||
| XM_011540400.2 | XP_011538702.1 | 3 | 1834 | |||
| XM_017016974.1 | XP_016872463.1 | 6 | 4254 | |||
| XM_017016975.1 | XP_016872464.1 | 5 | 4588 | |||
| XM_017016976.1 | XP_016872465.1 | 6 | 4529 | |||
| XM_017016977.1 | XP_016872466.1 | 6 | 2761 | |||
| XM_017016978.1 | XP_016872467.1 | 5 | 4642 | |||
| XM_017016979.1 | XP_016872468.1 | 4 | 2522 | |||
| XM_017016980.1 | XP_016872469.1 | 4 | 2484 | |||
| XM_017016981.1 | XP_016872470.1 | 5 | 4099 | |||
| XM_017016982.1 | XP_016872471.1 | 5 | 4115 | |||
| XM_017016983.1 | XP_016872472.1 | 5 | 4390 | |||
| XM_017016984.1 | XP_016872473.1 | 5 | 4374 | |||
| GenBank mRNA | AB011086.1 | - | 1 | 2538 | ||
| BC011672.2 | - | 3 | 1722 | 4300443 | ||

GPRIN2
Genomic Map