siRNA Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008511.2 | NP_001008511.2 | 2 | 330 | ||
| XM_006230755.1 | XP_006230817.1 | 2 | 443 | |||
| GenBank mRNA | AY280663.1 | - | 1 | 224 | ||
| BC104701.1 | - | 2 | 330 | |||

Esrra
Genomic Map