siRNA Details
Gene Symbol: Taar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008512.1 | NP_001008512.1 | 2 | 37 | ||
GenBank mRNA | AY702316.1 | - | 2 | 37 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
