siRNA Details
| Gene Symbol: RGD1310553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008517.1 | NP_001008517.1 | 2 | 143 | ||
| GenBank mRNA | BC086514.1 | - | 2 | 143 | 7134744 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310553| Gene Symbol: RGD1310553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008517.1 | NP_001008517.1 | 2 | 143 | ||
| GenBank mRNA | BC086514.1 | - | 2 | 143 | 7134744 | |