siRNA Details
| Gene Symbol: Abhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008520.1 | NP_001008520.1 | 6, 7 | 862 | ||
| GenBank mRNA | BC086351.1 | - | 6, 7 | 862 | 7302778 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd1| Gene Symbol: Abhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008520.1 | NP_001008520.1 | 6, 7 | 862 | ||
| GenBank mRNA | BC086351.1 | - | 6, 7 | 862 | 7302778 | |