siRNA Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008527.1 | NP_001008527.1 | 7 | 655 | ||
| GenBank mRNA | AY665586.1 | - | 7 | 655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aox3| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008527.1 | NP_001008527.1 | 7 | 655 | ||
| GenBank mRNA | AY665586.1 | - | 7 | 655 | ||