siRNA Details
| Gene Symbol: Slc10a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008555.1 | NP_001008555.1 | 3 | 1084 | ||
| GenBank mRNA | AY704415.1 | - | 3 | 1084 | ||
| AY825923.1 | - | 3 | 1084 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc10a4| Gene Symbol: Slc10a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008555.1 | NP_001008555.1 | 3 | 1084 | ||
| GenBank mRNA | AY704415.1 | - | 3 | 1084 | ||
| AY825923.1 | - | 3 | 1084 | |||