siRNA Details
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008722.1 | NP_001008722.1 | 2, 3 | 353 | ||
| XM_006255737.2 | XP_006255799.1 | 3, 4 | 401 | |||
| GenBank mRNA | AB120371.1 | - | 2, 3 | 353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irf8| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008722.1 | NP_001008722.1 | 2, 3 | 353 | ||
| XM_006255737.2 | XP_006255799.1 | 3, 4 | 401 | |||
| GenBank mRNA | AB120371.1 | - | 2, 3 | 353 | ||