siRNA Details
| Gene Symbol: Fga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008724.1 | NP_001008724.1 | 6 | 617 | ||
| NM_052797.1 | NP_434684.1 | 5 | 617 | |||
| XM_006232532.2 | XP_006232594.1 | 5 | 661 | |||
| XM_006232533.2 | XP_006232595.1 | 5 | 653 | |||
| GenBank mRNA | AY310161.1 | - | 6 | 617 | ||
| BC127465.1 | - | 5 | 617 | 8375711 | ||
| FQ209852.1 | - | 5 | 635 | |||

Fga
Genomic Map