siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008725.3 | NP_001008725.2 | 5 | 689 | ||
| XM_006231928.2 | XP_006231990.1 | 7 | 942 | |||
| XM_008760736.1 | XP_008758958.1 | 7 | 1787 | |||
| GenBank mRNA | AY310138.1 | - | 659 | |||
| JQ013730.1 | - | 5 | 1040 | |||
| M92340.1 | - | 5 | 723 | |||

Il6st
Genomic Map