siRNA Details
| Gene Symbol: Krt77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008807.1 | NP_001008807.1 | 5 | 1036 | ||
| XM_006242435.2 | XP_006242497.1 | 5 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt77| Gene Symbol: Krt77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008807.1 | NP_001008807.1 | 5 | 1036 | ||
| XM_006242435.2 | XP_006242497.1 | 5 | 1179 | |||