siRNA Details
| Gene Symbol: Krt73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008808.1 | NP_001008808.1 | 9 | 1414 | ||
| XM_008765703.1 | XP_008763925.1 | 9 | 1454 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt73| Gene Symbol: Krt73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008808.1 | NP_001008808.1 | 9 | 1414 | ||
| XM_008765703.1 | XP_008763925.1 | 9 | 1454 | |||