siRNA Details
| Gene Symbol: Kb23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008813.1 | NP_001008813.1 | 10 | 1613 | ||
| XM_006242350.2 | XP_006242412.1 | 9 | 1552 | |||
| GenBank mRNA | AB355639.1 | - | 9 | 1507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kb23| Gene Symbol: Kb23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008813.1 | NP_001008813.1 | 10 | 1613 | ||
| XM_006242350.2 | XP_006242412.1 | 9 | 1552 | |||
| GenBank mRNA | AB355639.1 | - | 9 | 1507 | ||