siRNA Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303627.1 | NP_001290556.1 | 2 | 216 | ||
| NM_001303628.1 | NP_001290557.1 | 2 | 499 | |||
| NM_014730.3 | NP_055545.1 | 2 | 499 | |||
| XM_011539031.1 | XP_011537333.1 | 2 | 4914 | |||
| XM_011539032.1 | XP_011537334.1 | 3 | 4602 | |||
| GenBank mRNA | BC000371.2 | - | 2 | 448 | 2819615 | |
| BC016297.2 | - | 2 | 450 | 3832682 | ||
| BG718506.1 | - | 2 | 504 | |||
| BI458950.1 | - | 2 | 218 | |||
| BU845446.1 | - | 477 | ||||
| D63486.1 | - | 2 | 454 | |||

MLEC
Genomic Map