siRNA Details
| Gene Symbol: Krt75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008828.1 | NP_001008828.1 | 1 | 340 | ||
| XM_006242325.2 | XP_006242387.1 | 1 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt75| Gene Symbol: Krt75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008828.1 | NP_001008828.1 | 1 | 340 | ||
| XM_006242325.2 | XP_006242387.1 | 1 | 540 | |||