siRNA Details
| Gene Symbol: RT1-CE15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008838.1 | NP_001008838.1 | 1 | 266 | ||
| Gene Symbol: LOC103694386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772782.1 | XP_008771004.1 | 2 | 431 | ||

RT1-CE15
Genomic Map