siRNA Details
| Gene Symbol: LOC362901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008877.2 | NP_001008877.1 | 6 | 743 | ||
| GenBank mRNA | AY310150.1 | - | 783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC362901| Gene Symbol: LOC362901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008877.2 | NP_001008877.1 | 6 | 743 | ||
| GenBank mRNA | AY310150.1 | - | 783 | |||