siRNA Details
Gene Symbol: LOC362921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008878.1 | NP_001008878.1 | 3, 4 | 188 | ||
GenBank mRNA | AY321343.1 | - | 3, 4 | 188 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC362921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008878.1 | NP_001008878.1 | 3, 4 | 188 | ||
GenBank mRNA | AY321343.1 | - | 3, 4 | 188 |