siRNA Details
| Gene Symbol: Krt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008899.1 | NP_001008899.1 | 1 | 548 | ||
| XM_006242436.2 | XP_006242498.1 | 1 | 1052 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt2| Gene Symbol: Krt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008899.1 | NP_001008899.1 | 1 | 548 | ||
| XM_006242436.2 | XP_006242498.1 | 1 | 1052 | |||