siRNA Details
| Gene Symbol: KIAA0586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244189.1 | NP_001231118.1 | 7 | 855 | ||
| NM_001244190.1 | NP_001231119.1 | 6 | 698 | |||
| NM_001244191.1 | NP_001231120.1 | 5 | 506 | |||
| NM_001244192.1 | NP_001231121.1 | 6 | 629 | |||
| NM_001244193.1 | NP_001231122.1 | 2 | 142 | |||
| NM_014749.3 | NP_055564.3 | 5 | 725 | |||
| GenBank mRNA | AB011158.1 | - | 6 | 681 | ||
| AK302579.1 | - | 2 | 144 | |||
| AK302635.1 | - | 5 | 465 | |||
| AK302718.1 | - | 2 | 147 | |||
| AK302836.1 | - | 7 | 855 | |||
| AK310356.1 | - | 2 | 158 | |||
| AY359881.1 | - | 6 | 578 | |||
| BC066647.1 | - | 5 | 680 | 6166031 | ||
| DC348119.1 | - | 5 | 348 | |||

KIAA0586
Genomic Map