siRNA Details
| Gene Symbol: Eci3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009275.1 | NP_001009275.1 | 9 | 919 | ||
| GenBank mRNA | BC088178.1 | - | 9 | 919 | 7324966 | |
| FQ209521.1 | - | 8 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eci3| Gene Symbol: Eci3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009275.1 | NP_001009275.1 | 9 | 919 | ||
| GenBank mRNA | BC088178.1 | - | 9 | 919 | 7324966 | |
| FQ209521.1 | - | 8 | 759 | |||