siRNA Details
| Gene Symbol: Ppdpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009316.1 | NP_001009316.1 | 657 | |||
| GenBank mRNA | BC086948.1 | - | 657 | 7323892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppdpf| Gene Symbol: Ppdpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009316.1 | NP_001009316.1 | 657 | |||
| GenBank mRNA | BC086948.1 | - | 657 | 7323892 | ||