siRNA Details
| Gene Symbol: Tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009344.2 | NP_001009344.2 | 5, 6 | 810 | ||
| GenBank mRNA | BC087142.1 | - | 5, 6 | 771 | 7127799 | |
| FQ219023.1 | - | 5, 6 | 742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbrg1| Gene Symbol: Tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009344.2 | NP_001009344.2 | 5, 6 | 810 | ||
| GenBank mRNA | BC087142.1 | - | 5, 6 | 771 | 7127799 | |
| FQ219023.1 | - | 5, 6 | 742 | |||