siRNA Details
| Gene Symbol: Mthfs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009349.1 | NP_001009349.1 | 3 | 2207 | ||
| XM_008766442.1 | XP_008764664.1 | 3 | 2353 | |||
| XM_008766443.1 | XP_008764665.1 | 3 | 3989 | |||
| XM_008766444.1 | XP_008764666.1 | 4 | 2424 | |||
| GenBank mRNA | BC087144.1 | - | 3 | 2207 | 7128624 | |

Mthfs
Genomic Map