siRNA Details
| Gene Symbol: Enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009391.1 | NP_001009391.1 | 6 | 1045 | ||
| XM_008770023.1 | XP_008768245.1 | 6 | 1095 | |||
| GenBank mRNA | BC087697.1 | - | 6 | 1045 | 7312419 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enoph1| Gene Symbol: Enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009391.1 | NP_001009391.1 | 6 | 1045 | ||
| XM_008770023.1 | XP_008768245.1 | 6 | 1095 | |||
| GenBank mRNA | BC087697.1 | - | 6 | 1045 | 7312419 | |