siRNA Details
| Gene Symbol: Nsdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009399.1 | NP_001009399.1 | 2 | 170 | ||
| XM_006229515.2 | XP_006229577.1 | 2 | 185 | |||
| GenBank mRNA | BC087626.1 | - | 2 | 170 | 7317527 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsdhl| Gene Symbol: Nsdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009399.1 | NP_001009399.1 | 2 | 170 | ||
| XM_006229515.2 | XP_006229577.1 | 2 | 185 | |||
| GenBank mRNA | BC087626.1 | - | 2 | 170 | 7317527 | |