siRNA Details
| Gene Symbol: Pomgnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009437.1 | NP_001009437.1 | 1 | 1325 | ||
| XM_008766681.1 | XP_008764903.1 | 2 | 1597 | |||
| XM_008766682.1 | XP_008764904.1 | 2 | 2962 | |||
| XM_008766683.1 | XP_008764905.1 | 3 | 1667 | |||
| GenBank mRNA | AJ868534.1 | - | 1 | 1325 | ||

Pomgnt2
Genomic Map