siRNA Details
| Gene Symbol: Ly49i9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009496.1 | NP_001009496.1 | 7 | 1298 | ||
| GenBank mRNA | AY649839.1 | - | 7 | 1298 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49i9| Gene Symbol: Ly49i9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009496.1 | NP_001009496.1 | 7 | 1298 | ||
| GenBank mRNA | AY649839.1 | - | 7 | 1298 | ||